missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromoethyl)phthalimide, 97%

Product Code. p-74264 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10646482

Brand: Thermo Scientific Acros 106970250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 574-98-1 | C10H8BrNO2 | 254.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-98-1
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.08
MDL Number MFCD00005902
InChI Key CHZXTOCAICMPQR-UHFFFAOYSA-N
Synonym n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione
PubChem CID 11325
IUPAC Name 2-(2-bromoethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 81.0°C to 84.0°C
Color Beige or Pink to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21,461
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents ca.10g/l,hardly soluble in ethanol and ether
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Bromoethyl)phthalimide, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.