missing translation for 'onlineSavingsMsg'
Learn More

N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione, 97%, Thermo Scientific™

Catalog Number p-7055407
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
885266-56-8
C22H24N2O4
380.444
MFCD03844710
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
24720887
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
This item is not returnable. View return policy
885266-56-8
C22H24N2O4
380.444
MFCD03844710
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
24720887
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 380.44
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'