missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Benzoyl-4-chlorophenyl)formamide, TRC

Product Code. 30329591
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329591 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329591 Supplier Toronto Research Chemicals Supplier No. TRCB207890250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Benzoyl-4-chlorophenyl)formamide
CAS 10352-28-0
Molecular Formula C14H10ClNO2
Quantity 250 mg
Synonym 2'-Benzoyl-4'-chloroformanilide, 5-Chloro-2-formamidobenzophenone, NSC 338521
InChI Formula InChI=1S/C14H10ClNO2/c15-11-6-7-13(16-9-17)12(8-11)14(18)10-4-2-1-3-5-10/h1-9H,(H,16,17)
SMILES Clc1ccc(NC=O)c(c1)C(=O)c2ccccc2
IUPAC Name N-(2-benzoyl-4-chlorophenyl)formamide
Molecular Weight (g/mol) 259.69
Formula Weight 259.04
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.