missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)pyrrolidine, 99%

Product Code. 10217543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217543 100 mL Glass bottle 100mL
10274460 5 mL Glass bottle 5mL
10244510 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10217543 Supplier Thermo Scientific Acros Supplier No. 103701000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7154-73-6 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7154-73-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00003182
InChI Key WRXNJTBODVGDRY-UHFFFAOYSA-N
Synonym 1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
PubChem CID 1344
IUPAC Name 2-pyrrolidin-1-ylethanamine
SMILES C1CCN(C1)CCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 0.9000g/mL
Boiling Point 66°C to 70°C (23.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4677 to 1.4697
Quantity 100 mL
Specific Gravity 0.9
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)pyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.