missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)piperidine, 98%

Product Code. 10265970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265970 5 mL Glass bottle 5mL
10636544 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265970 Supplier Thermo Scientific Acros Supplier No. 161610050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27578-60-5 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27578-60-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00006516
InChI Key CJNRGSHEMCMUOE-UHFFFAOYSA-N
Synonym n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine
PubChem CID 33944
IUPAC Name 2-piperidin-1-ylethanamine
SMILES C1CCN(CC1)CCN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 186°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4730 to 1.475
Quantity 5 mL
Beilstein 20, 67
Specific Gravity 0.89
Formula Weight 128.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)piperidine
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.