missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)glycine

Product Code. 16627052 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16627052

Brand: Thermo Scientific Acros 465870010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24123-14-6 | C4H10N2O2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector AMINOETHYLGLYCINE
CAS 24123-14-6
Melting Point 142°C to 147°C
Color White
Boiling Point 290°C
Flash Point 129°C
Infrared Spectrum Conforms
Assay Percent Range 96.0 to 104.0 % (Titration with NaOH)
Packaging glass bottle
Molecular Formula C4H10N2O2
MDL Number MFCD00144824
Quantity 1 g
InChI Key PIINGYXNCHTJTF-UHFFFAOYSA-N
SMILES NCCNCC(O)=O
IUPAC Name 2-[(2-aminoethyl)amino]acetic acid
Molecular Weight (g/mol) 118.14
Formula Weight 118.14
Physical Form Powder
Chemical Name or Material N-(2-Aminoethyl)glycine
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Aminoethyl)glycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N-(2-Aminoethyl)glycine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.