missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals

Catalog Number p-4089556
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1001-53-2
C4H10N2O
102.14
MFCD00008163
DAKZISABEDGGSV-UHFFFAOYSA-N
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
66082
N-(2-aminoethyl)acetamide
CC(=O)NCCN
This item is not returnable. View return policy
1001-53-2
C4H10N2O
102.14
MFCD00008163
DAKZISABEDGGSV-UHFFFAOYSA-N
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
66082
N-(2-aminoethyl)acetamide
CC(=O)NCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 50.0°C
Boiling Point 143.0°C (10.0 mmHg)
Flash Point >110°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)NH(CH2)2NH2
Quantity 5 g
Beilstein 04, III, 522
Formula Weight 102.14
Percent Purity 90%
Chemical Name or Material N-(2-Aminoethyl)acetamide
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Aminoethyl)acetamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'