missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride |
| CAS | 1391053-50-1 |
| Analyte or Component Names | N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H26 N2 O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride, Oxymetazoline Hydrochloride Imp. A (EP) as Hydrochloride, Benzeneacetamide, N-(2-aminoethyl)-4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethyl-, hydrochloride (1:1), Oxymetazoline Related Compound A as Hydrochloride, Oxymetazoline Hydrochloride Impurity A as Hydrochloride, Oxymetazoline Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C16H26N2O2.ClH/c1-10-8-13(16(3,4)5)15(20)11(2)12(10)9-14(19)18-7-6-17;/h8,20H,6-7,9,17H2,1-5H3,(H,18,19);1H |
| SMILES | Cl.Cc1cc(c(O)c(C)c1CC(=O)NCCN)C(C)(C)C |
| IUPAC Name | N-(2-aminoethyl)-2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction