missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride, Mikromol™

Product Code. 17902806
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17902806 100 mg 100mg
17900806 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17902806 Supplier Mikromol Supplier No. MM0065.03
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
CAS 10076-98-9
Analyte or Component Names N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H21 Br N2 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Bromhexine Hydrochloride Imp. D (EP), Bromhexine Imp. D (EP), N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride, Bromhexine Hydrochloride Imp. D (EP) as Dihydrochloride, Bromhexine Hydrochloride Impurity D as Dihydrochloride, Bromhexine Impurity D as Dihydrochloride
InChI Formula InChI=1S/C14H21BrN2.2ClH/c1-17(13-5-3-2-4-6-13)10-11-9-12(15)7-8-14(11)16;;/h7-9,13H,2-6,10,16H2,1H3;2*1H
SMILES Cl.Cl.CN(Cc1cc(Br)ccc1N)C2CCCCC2
IUPAC Name 4-bromo-2-[[cyclohexyl(methyl)amino]methyl]aniline;dihydrochloride
Molecular Weight (g/mol) 370.16
Quantity 100 mg
Formula Weight 368.0422 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.