missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride, Mikromol™

Product Code. 17900806
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17900806

Brand: Mikromol MM0065.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
CAS 10076-98-9
Analyte or Component Names N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H21 Br N2 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Bromhexine Hydrochloride Imp. D (EP), Bromhexine Imp. D (EP), N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride, Bromhexine Hydrochloride Imp. D (EP) as Dihydrochloride, Bromhexine Hydrochloride Impurity D as Dihydrochloride, Bromhexine Impurity D as Dihydrochloride
InChI Formula InChI=1S/C14H21BrN2.2ClH/c1-17(13-5-3-2-4-6-13)10-11-9-12(15)7-8-14(11)16;;/h7-9,13H,2-6,10,16H2,1H3;2*1H
SMILES Cl.Cl.CN(Cc1cc(Br)ccc1N)C2CCCCC2
IUPAC Name 4-bromo-2-[[cyclohexyl(methyl)amino]methyl]aniline;dihydrochloride
Molecular Weight (g/mol) 370.16
Quantity 25 mg
Formula Weight 368.0422 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.