missing translation for 'onlineSavingsMsg'
Learn More

N-2-Acetylguanine, 99%

Product Code. 10065863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065863 5 g Glass bottle 5g
10607811 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10065863 Supplier Thermo Scientific Acros Supplier No. 258400050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19962-37-9 | C7H7N5O2 | 193.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19962-37-9
Molecular Formula C7H7N5O2
Molecular Weight (g/mol) 193.17
MDL Number MFCD00078201
InChI Key MXSMRDDXWJSGMC-UHFFFAOYSA-N
Synonym 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide
PubChem CID 88319
IUPAC Name N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N=CN2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >260°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in acids and alkalies
Formula Weight 193.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-2-Acetylguanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.