missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dichlorophenyl)thiourea, 99%, Thermo Scientific Chemicals

Product Code. 11469276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469276 5 g 5g
11479276 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469276 Supplier Thermo Scientific Alfa Aesar Supplier No. A18117.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-91-6
Molecular Formula C7H6Cl2N2S
Molecular Weight (g/mol) 221.099
MDL Number MFCD00041156
InChI Key KUQHRGMPBWZVQR-UHFFFAOYSA-N
Synonym 1-2,6-dichlorophenyl thiourea, 2,6-dichlorophenyl thiourea, n-2,6-dichlorophenyl thiourea, thiourea, 2,6-dichlorophenyl, 1-2,6-dichlorophenyl-2-thiourea, amino 2,6-dichlorophenyl amino methane-1-thione, chembl29234, 2,6-dichlorophenyl-thiourea, thiourea,n-2,6-dichlorophenyl
PubChem CID 722776
IUPAC Name (2,6-dichlorophenyl)thiourea
SMILES C1=CC(=C(C(=C1)Cl)NC(=S)N)Cl

Specifications

Melting Point 155°C to 159°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2212144
Formula Weight 221.11
Percent Purity 99%
Chemical Name or Material N-(2,6-Dichlorophenyl)thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.