missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dichlorophenyl)thiourea, 98+%, Thermo Scientific™

Product Code. 15438939
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438939 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438939 Supplier Thermo Scientific Alfa Aesar Supplier No. L11314.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(2,4-Dichlorophenyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep
TRUSTED_SUSTAINABILITY

Specifications

CAS 6326-14-3
Melting Point 157°C to 160°C
Odor Odorless
Molecular Formula C7H6Cl2N2S
MDL Number MFCD00022161
Quantity 5 g
UN Number UN2811
Beilstein 2096917
Synonym 1-2,4-dichlorophenyl thiourea, n-2,4-dichlorophenyl thiourea, 2,4-dichlorophenyl thiourea, 1-2,4-dichlorophenyl-2-thiourea, 2,4-dichloro-phenyl-thiourea, thiourea, 2,4-dichlorophenyl, amino 2,4-dichlorophenyl amino methane-1-thione, dichlorophenylthiourea, maybridge1_008642, 2,4-dichlorophenylthiourae
Solubility Information Insoluble in water.
InChI Key HSWCDFOASWNGOM-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)NC(=S)N
IUPAC Name (2,4-dichlorophenyl)thiourea
Molecular Weight (g/mol) 221.099
PubChem CID 2746442
Formula Weight 221.11
Percent Purity ≥98%
Chemical Name or Material N-(2,4-Dichlorophenyl)thiourea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.