missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(4-Aminophenoxy)ethyl]-N,N-dimethylamine, 97%, Thermo Scientific™

Product Code. 10133533
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133533 250 mg Amber Glass Bottle 250mg
10577474 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133533 Supplier Thermo Scientific Maybridge Supplier No. CC51514CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62345-76-0
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.25
MDL Number MFCD07801177
InChI Key CCCVQPGAXZNTIL-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethoxy aniline, 4-2-dimethylamino-ethoxy-phenylamine, 4-2-dimethylaminoethoxy aniline, n-2-4-aminophenoxy ethyl-n,n-dimethylamine, benzenamine,4-2-dimethylamino ethoxy, 2-4-aminophenoxy ethyl dimethylamine, 4-2-dimethoxyaminoethoxy aniline, 4-2-dimethylaminoethyloxy aniline, 4-2-dimethylaminoethyloxy-aniline
PubChem CID 6484655
IUPAC Name 4-[2-(dimethylamino)ethoxy]aniline
SMILES CN(C)CCOC1=CC=C(N)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.25
Percent Purity 95%
Chemical Name or Material N-[2-(4-Aminophenoxy)ethyl]-N, N-dimethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.