missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4,6-Trimethylphenyl)thiourea, 99%, Thermo Scientific™

Product Code. 11499992
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499992 100 g 100g
11489992 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499992 Supplier Thermo Scientific Alfa Aesar Supplier No. A12083.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91059-97-1
Molecular Formula C10H14N2S
Molecular Weight (g/mol) 194.296
MDL Number MFCD00051488
InChI Key LBUGQLLKHAHCJJ-UHFFFAOYSA-N
Synonym n-2,4,6-trimethylphenyl thiourea, n-mesitylthiourea, 2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl-2-thiourea, 2,4,6-trimethyl-phenyl-thiourea, amino 2,4,6-trimethylphenyl amino methane-1-thione, 1-mesitylthiourea, acmc-20akda, maybridge1_006982
PubChem CID 2731118
IUPAC Name (2,4,6-trimethylphenyl)thiourea
SMILES CC1=CC(=C(C(=C1)C)NC(=S)N)C

Specifications

Melting Point 230°C to 231°C
Quantity 100 g
UN Number UN2811
Beilstein 2642667
Formula Weight 194.3
Percent Purity 99%
Chemical Name or Material N- (2,4,6-Trimethylphenyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.