Learn More
N-(2,3-Epoxypropyl)phthalimide, 95%, Thermo Scientific Chemicals
£99.60 - £190.00
Chemical Identifiers
CAS | 5455-98-1 |
---|---|
Molecular Formula | C11H9NO3 |
Molecular Weight (g/mol) | 203.20 |
MDL Number | MFCD00005896,MFCD04973349,MFCD04973350 |
InChI Key | DUILGEYLVHGSEE-UHFFFAOYNA-N |
Synonym | n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl |
PubChem CID | 21601 |
IUPAC Name | 2-(oxiran-2-ylmethyl)isoindole-1,3-dione |
SMILES | O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11955581
|
Thermo Scientific Acros
170040100 |
10 g | Glass Bottle |
£99.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965581
|
Thermo Scientific Acros
170040250 |
25 g | Glass bottle |
£190.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5455-98-1 | |
203.20 | |
DUILGEYLVHGSEE-UHFFFAOYNA-N | |
21601 | |
O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 |
C11H9NO3 | |
MFCD00005896,MFCD04973349,MFCD04973350 | |
n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl | |
2-(oxiran-2-ylmethyl)isoindole-1,3-dione |
Specifications
5455-98-1 | |
100.0 | |
White to Yellow | |
94% min. (GC) | |
MFCD00005896,MFCD04973349,MFCD04973350 | |
DUILGEYLVHGSEE-UHFFFAOYNA-N | |
2-(oxiran-2-ylmethyl)isoindole-1,3-dione | |
21601 | |
95% | |
N-(2, 3-Epoxypropyl)phthalimide |
94.0 | |
94.0°C to 100.0°C | |
Authentic | |
C11H9NO3 | |
n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl | |
O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 | |
203.20 | |
203.2 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes,on skin,or on clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 226-710-2
- N-(2,3-Epoxypropyl)phthalimide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only