missing translation for 'onlineSavingsMsg'
Learn More

N-(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)-n-methylamine hydrochloride, TRC

Product Code. 30386306
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386306 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386306 Supplier Toronto Research Chemicals Supplier No. TRCD45138550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)-n-methylamine hydrochloride
CAS 852227-85-1
Molecular Formula C10H14ClNO2
Quantity 50 mg
Synonym N-(2,3-Dihydro-1,4-Benzodioxin-6-Ylmethyl)-N-Methylamine, Hcl
InChI Formula InChI=1S/C10H13NO2.ClH/c1-11-7-8-2-3-9-10(6-8)13-5-4-12-9;/h2-3,6,11H,4-5,7H2,1H3;1H
SMILES Cl.CNCc1ccc2OCCOc2c1
IUPAC Name 1-(2,3-dihydro-1,4-benzodioxin-6-yl)-N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 215.68
Formula Weight 215.0713
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.