missing translation for 'onlineSavingsMsg'
Learn More

N-(2,3-Dichlorophenyl)piperazine Hydrochloride, TRC

Product Code. 30385945
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385945 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385945 Supplier Toronto Research Chemicals Supplier No. TRCD43575050MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorophenylpiperazine (hydrochloride)
CAS 119532-26-2
Molecular Formula C10 H12 Cl2 N2 . Cl H
Quantity 50 mg
Synonym 1-(2,3-Dichlorophenyl)piperazine Hydrochloride, Aripiprazole Related Compound C (USP), N-(2,3-Dichlorophenyl)piperazine Hydrochloride, Aripiprazole Imp. B (EP) as Hydrochloride
InChI Formula InChI=1S/C10H12Cl2N2.ClH/c11-8-2-1-3-9(10(8)12)14-6-4-13-5-7-14;/h1-3,13H,4-7H2;1H
SMILES Cl.Clc1cccc(N2CCNCC2)c1Cl
IUPAC Name 1-(2,3-dichlorophenyl)piperazine;hydrochloride
Molecular Weight (g/mol) 267.58
Formula Weight 266.0144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.