missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(2,3-Dichlorophenyl)-3,5-difluorobenzamide, 97%
Brand: Alfa Aesar™ H61301.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(2,3-Dichlorophenyl)-3,5-difluorobenzamide | |
302.11 | |
MFCD24848949 |
Specifications
N-(2,3-Dichlorophenyl)-3,5-difluorobenzamide | |
97% | |
MFCD24848949 |
Solid | |
302.11 | |
1g |