missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl]ethyl]-N'-methylurea, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl]ethyl]-N'-methylurea
Specifications
Specifications
| Chemical Name or Material | N-[2-[([2-[(Dimethylamino)methyl]thiazol-4-yl]methyl)sulfanyl]ethyl]-N'-methylurea |
| CAS | 82586-81-0 |
| Analyte or Component Names | N-[2-[([2-[(Dimethylamino)methyl]thiazol-4-yl]methyl)sulfanyl]ethyl]-N'-methylurea |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H20 N4 O S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Nizatidine Imp. I (EP), N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-N'-methylurea, N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methylurea, Nizatidine Impurity I |
| InChI Formula | InChI=1S/C11H20N4OS2/c1-12-11(16)13-4-5-17-7-9-8-18-10(14-9)6-15(2)3/h8H,4-7H2,1-3H3,(H2,12,13,16) |
| SMILES | CNC(=O)NCCSCc1csc(CN(C)C)n1 |
| IUPAC Name | 1-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-3-methylurea |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction