missing translation for 'onlineSavingsMsg'
Learn More

N-(2,2-Dimethoxyethyl)prop-2-enamide, TRC

Product Code. 30388369
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388369 1 g
30381832 500 mg
30380492 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388369 Supplier Toronto Research Chemicals Supplier No. TRCD4650901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49707-23-5
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.18
Synonym N-(2,2-Dimethoxyethyl)-2-propenamide, (2,2-Dimethoxyethyl)acrylamide, Acrylamidoacetaldehyde Dimethyl Acetal, N-Acryloylaminoacetaldehyde Dimethyl Acetal
IUPAC Name N-(2,2-dimethoxyethyl)prop-2-enamide
SMILES COC(CNC(=O)C=C)OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H13NO3/c1-4-6(9)8-5-7(10-2)11-3/h4,7H,1,5H2,2-3H3,(H,8,9)
Formula Weight 159.0895
Chemical Name or Material N-(2,2-Dimethoxyethyl)prop-2-enamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.