missing translation for 'onlineSavingsMsg'
Learn More

N-(2,2-Diethoxyethyl)-N-(2-thienylmethyl)amine, 97%, Thermo Scientific™

Product Code. 10130892
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10130892 1 g Amber Glass Bottle 1g
10283562 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10130892 Supplier Thermo Scientific Maybridge Supplier No. S09640DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 113825-05-1
Molecular Formula C11H19NO2S
Molecular Weight (g/mol) 229.338
MDL Number MFCD00173734
InChI Key YRCGLUZNPJCFOZ-UHFFFAOYSA-N
Synonym 2,2-diethoxy-n-thiophen-2-ylmethyl ethanamine, 2-thiophenemethanamine,n-2,2-diethoxyethyl, n-2,2-diethoxyethyl-n-2-thienylmethyl amine, 2,2-diethoxyethyl thiophen-2-ylmethyl amine, acmc-20mj4j, 2,2-diethoxyethyl 2-thienylmethyl amine, 2,2-diethoxy-n-thiophen-2-ylmethyl ethaneamine, 2,2-bis ethyloxy-n-2-thienylmethyl ethanamine, n-2,2-diethyoxyethyl-n-2-thienylmethyl amine
PubChem CID 2795396
IUPAC Name 2,2-diethoxy-N-(thiophen-2-ylmethyl)ethanamine
SMILES CCOC(CNCC1=CC=CS1)OCC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 229.34
Percent Purity 97%
Chemical Name or Material N-(2, 2-diethoxyethyl)-N-(2-thienylmethyl)amine
compliance-icons
Product Identifier
  • N-(2,2-Diethoxyethyl)-N-(2-thienylmethyl)amine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.