Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Product Code. 10186563 Shop All Macherey-Nagel Products
Click to view available options
Color:
Blue
Red
Silver
Septum:
Natural Rubber/Butyl/TEF
Natural rubber / Butyl red orange/ TEF colorless
PTFE/Silicone/PTFE
Silicone/PTFE
Silicone/PTFE, Pre-slit
Virgin PTFE
Color (Seal):
Red/Orange/Clear
Red/White/Red
White
White/Blue
White/Red
Thickness (Metric):
0.25 mm
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 100
Product Code. 10186563
Brand: Macherey-Nagel™ 70284
Only null left
Add to basket
Add to basket
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
  • Ready assembled crimp closures and plain crimp caps N 11 
  • Septa are assembled, caps are ready-to-use.
  • Septa are shown for illustration purposes only, images upside down.
  • Packs of 100 closures each
Specifications
Color (Seal) White
Color Silver
Material Aluminum, PTFE
Septum Virgin PTFE
Quantity 100 Pk.
Thickness (Metric) 0.25 mm