missing translation for 'onlineSavingsMsg'
Learn More

(+)-N-[1-(S)-Ethoxycarbonxyl-3-phenylpropyl]-L-alanine, TRC

Product Code. 30135772
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135772 1 g 1g
30008598 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135772 Supplier Toronto Research Chemicals Supplier No. TRCE8915861G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82717-96-2
Molecular Formula C15 H21 N O4
Molecular Weight (g/mol) 279.33
Synonym (2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid, Quinapril Hydrochloride Imp. B (EP), Ramipril Imp. F (EP), Enalapril Maleate Imp. B (EP), Benzenebutanoic acid, α-[[(1S)-1-carboxyethyl]amino]-, 1-ethyl ester, (αS)-, Benzenebutanoic acid, α-[(1-carboxyethyl)amino]-, monoethyl ester, [S-(R*,R*)]-, Benzenebutanoic acid, α-[[(1S)-1-carboxyethyl]amino]-, monoethyl ester, (αS)- (9CI), N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, N-[1(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
IUPAC Name (2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoic acid
SMILES CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)O

Specifications

InChI Formula InChI=1S/C15H21NO4/c1-3-20-15(19)13(16-11(2)14(17)18)10-9-12-7-5-4-6-8-12/h4-8,11,13,16H,3,9-10H2,1-2H3,(H,17,18)/t11-,13-/m0/s1
Quantity 1 g
Formula Weight 279.1471
Shipping Condition Room Temperature
Chemical Name or Material (2S)-2-([(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.