missing translation for 'onlineSavingsMsg'
Learn More

N-1-Naphthylethylenediamine Dihydrochloride, TRC

Product Code. 30473847
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473847 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473847 Supplier Toronto Research Chemicals Supplier No. TRCN37248010G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride
CAS 1465-25-4
Purity Grade Notes HPLC
Molecular Formula C12H16Cl2N2
Quantity 10 g
Synonym N1-1-Naphthalenyl-1,2-ethanediamine Hydrochloride (1:2), N-1-Naphthalenyl-1,2-ethanediamine Dihydrochloride, Bratton-Marshall Reagent, N-(1-Naphthyl)ethylendiamine Dihydrochloride, NEDA, NEDA (amine)
InChI Formula InChI=1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H
SMILES NCCNC1=C2C(C=CC=C2)=CC=C1
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 186.2523646
Formula Weight 258.0691
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.