missing translation for 'onlineSavingsMsg'
Learn More
Learn More

N-1-Naphthylethylene Diamine Dihydrochloride, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
5g
10g
Description
N-1-Naphthylethylene Diamine Dihydrochloride, Certified AR for Analysis
Specifications
Color | Beige |
Melting Point | 200°C |
Packaging | Glass Jar |
Quantity | 10g |
Formula Weight | 259.17g/mol |
Physical Form | Solid |
Safety and Handling

Product Identifier
- N-(1-Naphthyl)ethylenediamine, dihydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.