missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)thiourea, 97%, Thermo Scientific™

Product Code. 11416380
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416380 250 g 250g
11406380 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416380 Supplier Thermo Scientific Alfa Aesar Supplier No. A19024.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-88-4
Molecular Formula C11H10N2S
Molecular Weight (g/mol) 202.275
MDL Number MFCD00041824
InChI Key PIVQQUNOTICCSA-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
PubChem CID 736366
ChEBI CHEBI:82256
IUPAC Name naphthalen-1-ylthiourea
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=S)N

Specifications

Melting Point 185°C to 191°C
Odor Odorless
Quantity 250 g
UN Number UN1651
Beilstein 778118
Merck Index 14,722
Formula Weight 202.29
Percent Purity 97%
Chemical Name or Material N-(1-Naphthyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.