missing translation for 'onlineSavingsMsg'
Learn More

n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™

Product Code. 11423811 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423811 1 g 1g
17871371 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423811 Supplier MP Biomedicals Supplier No. 0215125201
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as an ion pairing reagent for HPLC and as an anionic surfactant. n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals has a low molecular weight that will not form micelles in solution.

Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.

  • The anionic sulfonate counterion permits the separation and resolution of positively charged analytes
  • Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value
  • For ion-pair chromatography
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C
Quantity 1 g
Molecular Formula C6H13NaO3S
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES [Na+].CCCCCCS([O-])(=O)=O
IUPAC Name sodium hexane-1-sulfonate
Molecular Weight (g/mol) 188.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.