missing translation for 'onlineSavingsMsg'
Learn More

N-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™

Product Code. 11456065 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456065 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456065 Supplier MP Biomedicals Supplier No. 0215125225
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as an ion pairing reagent for HPLC and as an anionic surfactant. N-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals is a low molecular weight alkylsulfonate. The anionic sulfonate counterion permits the separation and resolution of posit

Because of its low molecular weight, Sodium hexanesulfonate will not form micelles in solution and therefore has no CMC value. This product is used for ion-pair chromatography. It is also utilized in the analysis of small organic compounds, pharmaceutical products and metabolites by such techniques as HPLC.

Specifications

Form White Flakes
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.