missing translation for 'onlineSavingsMsg'
Learn More

N,1-Di-Boc-L-histidine dicyclohexylammonium salt, 98%

Product Code. 15447586
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447586 25 g 25g
15437586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447586 Supplier Thermo Scientific Alfa Aesar Supplier No. H62570.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31687-58-8 | C28H48N4O6 | 536.714 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31687-58-8
Molecular Formula C28H48N4O6
Molecular Weight (g/mol) 536.714
MDL Number MFCD06804675
InChI Key WBGMQHNUPJENDC-MERQFXBCSA-N
Synonym boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
PubChem CID 16211319
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Melting Point 160°C to 165°C (decomposition)
Quantity 25 g
Formula Weight 536.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N,1-Di-Boc-L-histidine dicyclohexylammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.