missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
IUPAC Name | 1-(1-benzothiophen-2-yl)-N-methylmethanamine;hydrochloride |
Formula Weight | 213.73 |
Percent Purity | 97% |
Chemical Name or Material | N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'