missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1,1-Dimethylethoxy)carbonyl Tranexamic Acid Dimer, TRC
Specifications
Specifications
| Chemical Name or Material | N-(1,1-Dimethylethoxy)carbonyl Tranexamic Acid Dimer |
| Analyte or Component Names | N-(1,1-Dimethylethoxy)carbonyl Tranexamic Acid Dimer |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H35 N O6 |
| Synonym | N-(1,1-Dimethylethoxy)carbonyl trans,trans-4,4’-(Iminodimethylene)di(cyclohexanecarboxylic) Acid |
| InChI Formula | InChI=1S/C21H35NO6/c1-21(2,3)28-20(27)22(12-14-4-8-16(9-5-14)18(23)24)13-15-6-10-17(11-7-15)19(25)26/h14-17H,4-13H2,1-3H3,(H,23,24)(H,25,26)/t14-,15-,16-,17- |
| Solution Type | Neat |
| SMILES | CC(C)(C)OC(=O)N(C[C@@H]1CC[C@H](CC1)C(=O)O)C[C@@H]2CC[C@H](CC2)C(=O)O |
| IUPAC Name | 4-[[(4-carboxycyclohexyl)methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]methyl]cyclohexane-1-carboxylic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?