missing translation for 'onlineSavingsMsg'
Learn More

N-(1,1-Dimethylethoxy)carbonyl N-(4-Hydroxymethylcyclohexyl)methyl-tranexamic Acid Methyl Ester, TRC

Product Code. 30385828
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385828 25 mg 25mg
30379398 100 mg 100mg
30384856 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385828 Supplier Toronto Research Chemicals Supplier No. TRCD47302525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(1,1-Dimethylethoxy)carbonyl N-(4-Hydroxymethylcyclohexyl)methyl-tranexamic Acid Methyl Ester
Analyte or Component Names N-(1,1-Dimethylethoxy)carbonyl N-(4-Hydroxymethylcyclohexyl)methyl-tranexamic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H39 N O5
Synonym N-(1,1-Dimethylethoxy)carbonyl N-(4-Hydroxymethylcyclohexyl)methyl-tranexamic Acid Methyl Ester
InChI Formula InChI=1S/C22H39NO5/c1-22(2,3)28-21(26)23(13-16-5-7-18(15-24)8-6-16)14-17-9-11-19(12-10-17)20(25)27-4/h16-19,24H,5-15H2,1-4H3/t16-,17-,18-,19-
Solution Type Neat
SMILES COC(=O)[C@@H]1CC[C@@H](CN(C[C@@H]2CC[C@@H](CO)CC2)C(=O)OC(C)(C)C)CC1
IUPAC Name methyl 4-[[[4-(hydroxymethyl)cyclohexyl]methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]methyl]cyclohexane-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 397.549
Quantity 25 mg
Formula Weight 397.283
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.