missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106478
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MZF-1.
Binds to target promoter DNA and functions as transcription regulator. Regulates transcription from the PADI1 and CDH2 promoter. May be one regulator of transcriptional events during hemopoietic development. [UniProt].
Specifications
| MZF1 | |
| Polyclonal | |
| Unconjugated | |
| MZF1 | |
| myeloid zinc finger 1; Myeloid zinc finger protein 1; Myeloid zinc finger protein 2; myeloid-specific retinoic acid-responsive zinc finger protein; MZF; MZF1; MZF-1; MZF1B; Mzf2; Mzf-2; Zfp121; Zfp98; Zinc finger and SCAN domain-containing protein 6; zinc finger protein 121; Zinc finger protein 42; zinc finger protein 42 (myeloid-specific retinoic acid-responsive); zinc finger protein 98; Znf42; ZSCAN6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 109889, 361508, 7593 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28698 | |
| MZF1 | |
| A synthesized peptide derived from human MZF1(Accession P28698), corresponding to amino acid residues M1-P40. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction