missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MZB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584122
This item is not returnable.
View return policy
Description
Immunogen sequence: TATAPQLDDE EMYSAHMPAH LRCDACRAVA YQMWQNLAKA ETKLHTSNSG GRRELSELVY TDVLDRS Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 72%.
MZB1 associates with immunoglobulin M (IgM) heavy and light chains and promotes IgM assembly and secretion. May exert its effect by acting as a molecular chaperone or as an oxidoreductase as it displays a low level of oxidoreductase activity. Isoform 2 may be involved in regulation of apoptosis. Helps to diversify peripheral B-cell functions by regulating Ca(2+) stores, antibody secretion and integrin activation.; Acts as a hormone-regulated adipokine/proinflammatory cytokine that is implicated in causing chronic inflammation, affecting cellular expansion and blunting insulin response in adipocytes. May have a role in the onset of insulin resistance.
Specifications
| MZB1 | |
| Polyclonal | |
| Unconjugated | |
| MZB1 | |
| caspase-2 binding protein; HSPC190; marginal zone B and B1 cell specific protein; marginal zone B and B1 cell-specific protein; marginal zone B- and B1-cell-specific protein; MEDA7; MEDA-7; Mesenteric estrogen-dependent adipose 7; mesenteric oestrogen-dependent adipose gene- 7; MGC29506; MZB1; PACAP; pERp1; plasma cell-induced ER protein 1; plasma cell-induced resident endoplasmic reticulum protein; Plasma cell-induced resident ER protein; proapoptotic caspase adapter protein; proapoptotic caspase adaptor protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51237 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WU39 | |
| MZB1 | |
| Recombinant protein corresponding to Human MZB1. Recombinant protein control fragment (Product #RP-104951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction