missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST3 Antibody (127A) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50516-0.025ml
This item is not returnable.
View return policy
Description
MYST3 Monoclonal antibody specifically detects MYST3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| MYST3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| EC 2.3.1.48, EC 3.1.26.4, histone acetyltransferase MYST3, KAT6A, MGC167033, MOZMonocytic leukemia zinc finger protein, MYST histone acetyltransferase (monocytic leukemia) 3, MYST-3, runt-related transcription factor binding protein 2, Runt-related transcription factor-binding protein 2, RUNXBP2, Zinc finger protein 220, ZNF220MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3 | |
| GST-MYST3 (172-321aa) recombinant protein human MYST3 | |
| 0.025 mL | |
| Epigenetics | |
| 7994 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG3 κ |
| Western Blot, Immunoprecipitation | |
| 127A | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction