missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYST2 Monoclonal Antibody (1D9H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538549
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| MYST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O95251, Q5SVQ0 | |
| Kat7 | |
| Purified recombinant fragment of human KAT7 (aa: 1-200) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1D9H9 | |
| Unconjugated | |
| Kat7 | |
| EC 2.3.1.48; HBO1; HBOa; histone acetyltransferase binding to ORC1; Histone acetyltransferase KAT7; histone acetyltransferase MYST2; K (lysine) acetyltransferase 7; K(lysine) acetyltransferase 7; Kat7; Lysine acetyltransferase 7; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2; MYST histone acetyltransferase 2; MYST protein 2; MYST2; MYST-2; ZC2HC7 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11143, 217127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction