missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYSM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584747
This item is not returnable.
View return policy
Description
Immunogen sequence: VNCIGRIHTY LELIGAINFG CEQAVYNRPQ TVDKVRIRDR KDAVEAYQLA QRLQSMRTRR RRVRDPWGNW CDAKDLEGQT FEHLS Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
Metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Preferentially deubiquitinates monoubiquitinated H2A in hyperacetylated nucleosomes. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. Acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation.
Specifications
| MYSM1 | |
| Polyclonal | |
| Unconjugated | |
| Mysm1 | |
| 2ADUB; 2A-DUB; C130067A03Rik; C530050H10Rik; Histone H2A deubiquitinase MYSM1; KIAA1915; Myb like, SWIRM and MPN domains 1; myb-like, SWIRM and MPN domain-containing protein 1; myb-like, SWIRM and MPN domains 1; Mysm1; RGD1311787; RP4-592A1.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VVJ2 | |
| Mysm1 | |
| Recombinant protein corresponding to Human MYSM1. Recombinant protein control fragment (Product #RP-105418). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction