missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYSM1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95199-0.02ml
This item is not returnable.
View return policy
Description
MYSM1 Polyclonal antibody specifically detects MYSM1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MYSM1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp779J1554, DKFZp779J1721,2A-DUB, histone H2A deubiquitinase MYSM1, KIAA1915, Myb-like, SWIRM and MPN domains 1,2ADUB, RP4-592A1.1, SWIRM and MPN domain-containing protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 720-821 of human MYSM1 (NP_001078956.1). SYRLPYKFEVQQMLEEPQWGLVFEKTRWIIEKYRLSHSSVPMDKIFRRDSDLTCLQKLLECMRKTLSKVTNCFMAEEFLTEIENLFLSNYKSNQENGVTEEN | |
| 0.02 mL | |
| Cell Biology | |
| 114803 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction