missing translation for 'onlineSavingsMsg'
Learn More

(-)-Myrtenal, 98%

Product Code. 10296780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296780 10 g Glass bottle 10g
10792241 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296780 Supplier Thermo Scientific Acros Supplier No. 203620100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18486-69-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18486-69-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00074768
InChI Key KMRMUZKLFIEVAO-UHFFFAOYNA-N
Synonym --myrtenal, 1r-2-pinen-10-al, fema no. 3395, 1r-6,6-dimethylbicyclo 3.1.1 hept-2-en-2-carboxaldehyde, 2-norpinene-2-carboxaldehyde, 6,6-dimethyl-8ci, bicyclo 3.1.1 hept-2-ene-2-carboxaldehyde, 6,6-dimethyl-, 1s, 1r---myrtenal, 1r---myrtenal, analytical standard, --myrtenal sum of enantiomers, gc
PubChem CID 1201529
SMILES CC1(C)C2CC1C(C=O)=CC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 220.0°C to 221.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5029 to 1.5049
Quantity 10 g
Beilstein 07, 161
Specific Gravity 0.98
Specific Rotation Condition −15° (22°C neat)
Specific Rotation − 15.00
IUPAC Name 6,6-dimethylbicyclo[3.1.1]hept-2-ene-2-carbaldehyde
Formula Weight 150.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (-)-Myrtenal, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.