missing translation for 'onlineSavingsMsg'
Learn More

(-)-Myrtenal, 98%, Thermo Scientific Chemicals

Catalog Number p-169912
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18486-69-6
C10H14O
150.22
MFCD00074768
KMRMUZKLFIEVAO-UHFFFAOYNA-N
--myrtenal, 1r-2-pinen-10-al, fema no. 3395, 1r-6,6-dimethylbicyclo 3.1.1 hept-2-en-2-carboxaldehyde, 2-norpinene-2-carboxaldehyde, 6,6-dimethyl-8ci, bicyclo 3.1.1 hept-2-ene-2-carboxaldehyde, 6,6-dimethyl-, 1s, 1r---myrtenal, 1r---myrtenal, analytical standard, --myrtenal sum of enantiomers, gc
1201529
CC1(C)C2CC1C(C=O)=CC2
This item is not returnable. View return policy
18486-69-6
C10H14O
150.22
MFCD00074768
KMRMUZKLFIEVAO-UHFFFAOYNA-N
--myrtenal, 1r-2-pinen-10-al, fema no. 3395, 1r-6,6-dimethylbicyclo 3.1.1 hept-2-en-2-carboxaldehyde, 2-norpinene-2-carboxaldehyde, 6,6-dimethyl-8ci, bicyclo 3.1.1 hept-2-ene-2-carboxaldehyde, 6,6-dimethyl-, 1s, 1r---myrtenal, 1r---myrtenal, analytical standard, --myrtenal sum of enantiomers, gc
1201529
CC1(C)C2CC1C(C=O)=CC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 220.0°C to 221.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5029 to 1.5049
Quantity 10 g
Beilstein 07, 161
Specific Gravity 0.98
Specific Rotation Condition −15° (22°C neat)
Specific Rotation − 15.00
IUPAC Name 6,6-dimethylbicyclo[3.1.1]hept-2-ene-2-carbaldehyde
Formula Weight 150.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (-)-Myrtenal, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'