missing translation for 'onlineSavingsMsg'
Learn More

Myristyltrimethylammonium Bromide, TRC

Product Code. 30136788
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136788 25 g 25g
30135686 100 g 100g
17931375 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136788 Supplier Toronto Research Chemicals Supplier No. TRCM88504025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17 H38 N . Br
Molecular Weight (g/mol) 336.39
Synonym 1-Tetradecanaminium, N,N,N-trimethyl-, bromide (9CI), Ammonium, trimethyltetradecyl-, bromide (8CI), Trimethyltetradecylammonium bromide (6CI,7CI), (1-Tetradecyl)trimethylammonium bromide, C14TAB, Cetrimide BP, MTAB, Microcide II, Morpan T, Myristyltrimethylammonium bromide, Myrtrimonium bromide, Mytab, N,N,N-Trimethyl-1-tetradecanaminium bromide, N-Tetradecyl-N,N,N-trimethylammonium bromide, Pentonium 4Br40, Quaternium 13, Querton 14Br40, TTAB, TTAB (surfactant), Tetradecyltrimethylammonium bromide, Tetradonium bromide, Trimethylmyristylammonium bromide, n-Tetradecyltrimethylammonium bromide
IUPAC Name trimethyl(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1
Formula Weight 335.2188
Percent Purity >95%
Chemical Name or Material Cetrimide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.