missing translation for 'onlineSavingsMsg'
Learn More

Myristyltrimethylammonium bromide, 99%

Product Code. 10582821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582821 500 g Plastic bottle 500g
10439930 5 g Glass bottle 5g
10266570 100 g Plastic bottle 100g
10501794 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10582821 Supplier Thermo Scientific Acros Supplier No. 229375000
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
PubChem CID 14250
ChEBI CHEBI:3565
IUPAC Name trimethyl(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 245°C to 250°C
Color White
pH 6 to 8 (1% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Plastic bottle
Linear Formula CH3(CH2)13N(CH3)3Br
Quantity 500 g
Merck Index 15,6422
Solubility Information Solubility in water: 100g/L (20°C)
Formula Weight 336.39
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Myristyltrimethylammonium bromide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Myristyltrimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.