missing translation for 'onlineSavingsMsg'
Learn More

Myristic Acid, TRC

Product Code. 17959264
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959264 5 g 5g
17978024 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17959264 Supplier Toronto Research Chemicals Supplier No. TRCM8846105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14 H28 O2
Molecular Weight (g/mol) 228.37
Synonym Tetradecanoic Acid, Myristic acid (8CI), 1-Tetradecanoic acid, 1-Tridecanecarboxylic acid, Edenor C 14, Edenor C 14/98-100, Edenor C14-99, Edenor C14-99 (C), Emery 655, Hystrene 9014, Imex C 1499, Kortacid 1499, Lunac M 98, Lunac MY 98, MY 98, Myristates, NAA 104, NAA 142, NSC 5028, Neo-Fat 14, POFAC 1498, Palmac 98-14, Philacid 1400, Prifac 2942, Prifrac 2942, Sinar FA 1499, Univol U 316S, n-Tetradecan-1-oic acid, n-Tetradecanoic acid, n-Tetradecoic acid
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)
Formula Weight 228.2089
Chemical Name or Material Myristic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.