missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Myristic acid, 99%

Product Code. 10143110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143110 1 kg Glass bottle 1kg
10792054 100 g Glass bottle 100g
10509253 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143110 Supplier Thermo Scientific Chemicals Supplier No. 156960010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 57.0°C
Color White
Boiling Point 326.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)12CO2H
Quantity 1 kg
Beilstein 02, 365
Merck Index 15, 6419
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, methanol, ether, petr.ether,, benzene and chloroform
Formula Weight 228.37
Percent Purity 99%
Physical Form Crystalline Mass, Powder, Flakes or Chunks
Chemical Name or Material Myristic acid
Show More Show Less
compliance-icons
Product Identifier
  • Myristic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.