missing translation for 'onlineSavingsMsg'
Learn More

MYRIP Antibody, Novus Biologicals™

Product Code. 18407810 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407810 0.1 mL 0.1mL
18456710 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18407810 Supplier Novus Biologicals Supplier No. NBP184137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYRIP Polyclonal antibody specifically detects MYRIP in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYRIP
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: SALPSWKSVDRLDETNLAPVLQSPDGNWVALKDGAPPPTRLLAKPKSGTFQALEVASSVASAYDEMGSDSEEDFDWSEALSKLCPRSRALPRNPQPQPTQAQSSDQGPIAASPSSALSPNPEAMCSDSETSSAGSSR
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25924
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.