missing translation for 'onlineSavingsMsg'
Learn More

Myricetin, MedChemExpress

Product Code. 16482427
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482427 50 mg 50mg
16431756 500 mg 500mg
16426636 100 mg 100mg
16401166 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482427 Supplier Medchem Express Supplier No. HY15097
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Myricetin is a common plant-derived flavonoid with a wide range of activities including strong anti-oxidant, anticancer, antidiabetic and anti-inflammatory activities.

Myricetin is a common plant-derived flavonoid with a wide range of activities including strong anti-oxidant, anticancer, antidiabetic and anti-inflammatory activities.

Specifications

Chemical Name or Material Myricetin
CAS 529-44-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O8
Quantity 50 mg
Synonym Cannabiscetin
Solubility Information DMSO : ≥ 31 mg/mL (97.41 mM) ∣Ethanol : 28.57 mg/mL (89.78 mM; Need ultrasonic)
SMILES OC1=CC(O)=C(C(C(O)=C(C2=CC(O)=C(O)C(O)=C2)O3)=O)C3=C1
Molecular Weight (g/mol) 318.24
Formula Weight 318.24
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.