missing translation for 'onlineSavingsMsg'
Learn More

Myrcene, 90%, tech., stabilized

Product Code. 10190370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190370 2.5 L Glass bottle 2.5 L
10448670 5 mL Glass bottle 5mL
10522911 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190370 Supplier Thermo Scientific Acros Supplier No. 128080025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-35-3 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-35-3 , 127-91-3 , 138-86-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
InChI Key UAHWPYUMFXYFJY-UHFFFAOYSA-N
Synonym myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
PubChem CID 31253
ChEBI CHEBI:17221
IUPAC Name 7-methyl-3-methylideneocta-1,6-diene
SMILES CC(=CCCC(=C)C=C)C

Specifications

Color Colorless to Yellow
Density 0.7950g/mL
Boiling Point 165.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
Refractive Index 1.4660 to 1.4720
Quantity 2.5 L
Beilstein 01,264
Merck Index 15,6416
Specific Gravity 0.795
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroform, ether and, glacial acetic acid
Stabilizer Stabilized with 0.01% alpha-Tocopherol
Formula Weight 136.24
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Myrcene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Myrcene, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.