missing translation for 'onlineSavingsMsg'
Learn More

Novex™ MYR PKC Zeta Pseudosubstrate

Product Code. 10717873
Click to view available options
Quantity:
1 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10717873

missing translation for 'mfr': Novex™ 77749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

MYR PKC Zeta Pseudosubstrate

Selective protein kinase C (PKC) zeta inhibitor.

Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.

Molecular Weight: 1928
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Form Lyophilized
Quantity 1 mg
Gene ID (Entrez) 5590
Inhibitor Properties Peptide
Inhibitors Kinase Inhibitor (PKC Inhibitor), Kinase Inhibitor
Molecular Weight (g/mol) 1928 Da
Purification Method HPLC
Shipping Condition Dry Ice
Specific Inhibit or Target PKCζ

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.