missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYPN Polyclonal antibody specifically detects MYPN in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MYPN |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 145 kDa (MYOP), MYOP145 kDa sarcomeric protein, myopalladin |
| Host Species | Rabbit |
| Immunogen | myopalladin. (Uniprot# Q86TC9) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?